siRNA Details
Gene Symbol: Tmem171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013903.1 | NP_001013925.1 | 2 | 248 | ||
XM_006231833.2 | XP_006231895.1 | 2 | 1165 | |||
XM_006231834.2 | XP_006231896.1 | 2 | 1164 | |||
XM_008760660.1 | XP_008758882.1 | 2 | 681 | |||
GenBank mRNA | AY185507.1 | - | 2 | 248 |
