siRNA Details
| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013910.2 | NP_001013932.1 | 7 | 1465 | ||
| GenBank mRNA | AF022085.1 | - | 431 | |||
| BC097370.1 | - | 7 | 1465 | 7444022 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnb4| Gene Symbol: Gnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013910.2 | NP_001013932.1 | 7 | 1465 | ||
| GenBank mRNA | AF022085.1 | - | 431 | |||
| BC097370.1 | - | 7 | 1465 | 7444022 | ||