siRNA Details
Gene Symbol: Ift80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013911.1 | NP_001013933.1 | 7 | 698 | ||
GenBank mRNA | BC081931.1 | - | 7 | 698 | 7113994 | |
BC081937.1 | - | 7 | 698 | 7115382 |
