siRNA Details
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013913.1 | NP_001013935.1 | 9 | 1715 | ||
XM_008761389.1 | XP_008759611.1 | 8 | 1359 | |||
XM_008761390.1 | XP_008759612.1 | 9 | 1899 | |||
GenBank mRNA | BC085920.1 | - | 9 | 1715 | 7108742 |
