siRNA Details
| Gene Symbol: Dtwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013921.1 | NP_001013943.1 | 3 | 923 | ||
| XM_006234910.2 | XP_006234972.1 | 4 | 902 | |||
| XM_006234914.2 | XP_006234976.1 | 4 | 701 | |||
| XM_006234915.2 | XP_006234977.1 | 4 | 836 | |||
| XM_008762154.1 | XP_008760376.1 | 4 | 1714 | |||
| GenBank mRNA | BC079068.1 | - | 3 | 923 | 7121624 | |

Dtwd1
Genomic Map