siRNA Details
| Gene Symbol: Dtwd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013921.1 | NP_001013943.1 | 4 | 1458 | ||
| XM_006234910.2 | XP_006234972.1 | 5 | 1437 | |||
| XM_006234914.2 | XP_006234976.1 | 5 | 1236 | |||
| XM_006234915.2 | XP_006234977.1 | 5 | 1371 | |||
| XM_008762154.1 | XP_008760376.1 | 5 | 2249 | |||
| GenBank mRNA | BC079068.1 | - | 4 | 1458 | 7121624 | |

Dtwd1
Genomic Map