siRNA Details
Gene Symbol: RGD1307752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013922.1 | NP_001013944.1 | 3 | 469 | ||
GenBank mRNA | BC089919.1 | - | 3 | 469 | 7306699 | |
FQ214713.1 | - | 3 | 462 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1307752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013922.1 | NP_001013944.1 | 3 | 469 | ||
GenBank mRNA | BC089919.1 | - | 3 | 469 | 7306699 | |
FQ214713.1 | - | 3 | 462 |