siRNA Details
| Gene Symbol: Ndrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013923.1 | NP_001013945.1 | 16 | 1589 | ||
| GenBank mRNA | BC078946.1 | - | 16 | 1589 | 7109582 | |
| BC091308.1 | - | 10 | 1278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndrg3| Gene Symbol: Ndrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013923.1 | NP_001013945.1 | 16 | 1589 | ||
| GenBank mRNA | BC078946.1 | - | 16 | 1589 | 7109582 | |
| BC091308.1 | - | 10 | 1278 | |||