siRNA Details
| Gene Symbol: Mtg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013924.1 | NP_001013946.1 | 7 | 945 | ||
| XM_006235817.2 | XP_006235879.1 | 7 | 1371 | |||
| XM_006235818.2 | XP_006235880.1 | 7 | 991 | |||
| XM_006235820.2 | XP_006235882.1 | 7 | 999 | |||
| XM_008762485.1 | XP_008760707.1 | 6 | 2505 | |||
| XM_008762486.1 | XP_008760708.1 | 7 | 1375 | |||
| XM_008762487.1 | XP_008760709.1 | 8 | 1092 | |||
| XM_008762488.1 | XP_008760710.1 | 8 | 1115 | |||
| XM_008762489.1 | XP_008760711.1 | 7 | 1014 | |||
| XM_008762490.1 | XP_008760712.1 | 8 | 1071 | |||
| XM_008762491.1 | XP_008760713.1 | 7 | 995 | |||
| XM_008762492.1 | XP_008760714.1 | 5 | 2286 | |||
| XM_008762493.1 | XP_008760715.1 | 7 | 1022 | |||
| XM_008762494.1 | XP_008760716.1 | 6 | 2505 | |||
| GenBank mRNA | BC083707.1 | - | 7 | 945 | 7190542 | |

Mtg2
Genomic Map