siRNA Details
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013946.1 | NP_001013968.1 | 10, 11 | 1394 | ||
XM_006241054.1 | XP_006241116.1 | 11, 12 | 1302 | |||
XM_006241055.1 | XP_006241117.1 | 10, 11 | 1404 | |||
GenBank mRNA | BC085785.1 | - | 10, 11 | 1394 | 7108413 |
