siRNA Details
| Gene Symbol: Akap8l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013946.1 | NP_001013968.1 | 10 | 1316 | ||
| XM_006241054.1 | XP_006241116.1 | 11 | 1224 | |||
| XM_006241055.1 | XP_006241117.1 | 10 | 1326 | |||
| GenBank mRNA | BC085785.1 | - | 10 | 1316 | 7108413 | |

Akap8l
Genomic Map