siRNA Details
| Gene Symbol: Ankrd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013948.1 | NP_001013970.1 | 4 | 615 | ||
| XM_006241530.2 | XP_006241592.1 | 4 | 886 | |||
| XM_006241531.2 | XP_006241593.1 | 3 | 518 | |||
| XM_006241532.2 | XP_006241594.1 | 3 | 2259 | |||
| XM_006241533.2 | XP_006241595.1 | 3 | 2185 | |||
| GenBank mRNA | AB095364.1 | - | 4 | 615 | ||
| BC105616.1 | - | 4 | 611 | 7453107 | ||

Ankrd46
Genomic Map