siRNA Details
| Gene Symbol: LOC300308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013952.1 | NP_001013974.1 | 1346 | |||
| GenBank mRNA | BC090074.1 | - | 1346 | 7114534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC300308| Gene Symbol: LOC300308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013952.1 | NP_001013974.1 | 1346 | |||
| GenBank mRNA | BC090074.1 | - | 1346 | 7114534 | ||