siRNA Details
| Gene Symbol: LOC300308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013952.1 | NP_001013974.1 | 3242 | |||
| GenBank mRNA | BC090074.1 | - | 3242 | 7114534 | ||
| Gene Symbol: RGD1566251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765279.1 | XP_008763501.1 | 4, 5 | 7481 | ||

LOC300308
Genomic Map