siRNA Details
| Gene Symbol: Pqbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013957.1 | NP_001013979.1 | 4, 5 | 431 | ||
| XM_006256707.1 | XP_006256769.1 | 4, 5 | 405 | |||
| XM_006256708.2 | XP_006256770.1 | 3, 4 | 306 | |||
| GenBank mRNA | BC059163.1 | - | 4, 5 | 431 | 6889015 | |
| FQ215363.1 | - | 1, 2 | 17 | |||
| FQ215670.1 | - | 1, 2 | 17 | |||
| FQ227645.1 | - | 1, 2 | 17 | |||

Pqbp1
Genomic Map