siRNA Details
| Gene Symbol: Pqbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013957.1 | NP_001013979.1 | 3 | 246 | ||
| XM_006256707.1 | XP_006256769.1 | 3 | 220 | |||
| XM_006256708.2 | XP_006256770.1 | 2 | 121 | |||
| GenBank mRNA | BC059163.1 | - | 3 | 246 | 6889015 | |

Pqbp1
Genomic Map