siRNA Details
| Gene Symbol: LOC302576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013959.1 | NP_001013981.1 | 1 | 762 | ||
| GenBank mRNA | BC083583.1 | - | 1 | 762 | 7131068 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC302576| Gene Symbol: LOC302576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013959.1 | NP_001013981.1 | 1 | 762 | ||
| GenBank mRNA | BC083583.1 | - | 1 | 762 | 7131068 | |