siRNA Details
| Gene Symbol: Mmgt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013967.1 | NP_001013989.1 | 2 | 1461 | ||
| GenBank mRNA | BC061881.1 | - | 2 | 1461 | 5599256 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmgt2| Gene Symbol: Mmgt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013967.1 | NP_001013989.1 | 2 | 1461 | ||
| GenBank mRNA | BC061881.1 | - | 2 | 1461 | 5599256 | |