siRNA Details
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013967.1 | NP_001013989.1 | 2 | 1061 | ||
GenBank mRNA | BC061881.1 | - | 2 | 1061 | 5599256 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013967.1 | NP_001013989.1 | 2 | 1061 | ||
GenBank mRNA | BC061881.1 | - | 2 | 1061 | 5599256 |