siRNA Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013973.1 | NP_001013995.1 | 5 | 552 | ||
| XM_006247579.2 | XP_006247641.1 | 5 | 720 | |||
| XM_006247580.2 | XP_006247642.1 | 6 | 839 | |||
| XM_006247585.2 | XP_006247647.1 | 5 | 697 | |||
| GenBank mRNA | AB552845.1 | - | 3 | 297 | ||
| AB552846.1 | - | 3 | 297 | |||
| BC083567.1 | - | 5 | 552 | 7112851 | ||

March10
Genomic Map