siRNA Details
| Gene Symbol: Ogfod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013976.1 | NP_001013998.1 | 9 | 1125 | ||
| GenBank mRNA | BC088238.1 | - | 9 | 1125 | 7326036 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfod3| Gene Symbol: Ogfod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013976.1 | NP_001013998.1 | 9 | 1125 | ||
| GenBank mRNA | BC088238.1 | - | 9 | 1125 | 7326036 | |