siRNA Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014762.1 | NP_001014762.1 | 5 | 366 | ||
| GenBank mRNA | BC082021.1 | - | 5 | 366 | 7129610 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdrg1| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014762.1 | NP_001014762.1 | 5 | 366 | ||
| GenBank mRNA | BC082021.1 | - | 5 | 366 | 7129610 | |