siRNA Details
| Gene Symbol: Tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017444.1 | NP_001017444.1 | 4 | 969 | ||
| GenBank mRNA | BC093382.1 | - | 4 | 969 | 7389050 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm2d2| Gene Symbol: Tm2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017444.1 | NP_001017444.1 | 4 | 969 | ||
| GenBank mRNA | BC093382.1 | - | 4 | 969 | 7389050 | |