siRNA Details
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017444.1 | NP_001017444.1 | 4 | 1074 | ||
GenBank mRNA | BC093382.1 | - | 4 | 1074 | 7389050 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017444.1 | NP_001017444.1 | 4 | 1074 | ||
GenBank mRNA | BC093382.1 | - | 4 | 1074 | 7389050 |