siRNA Details
| Gene Symbol: Faf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017445.1 | NP_001017445.1 | 7 | 539 | ||
| XM_006253617.2 | XP_006253679.1 | 6 | 564 | |||
| XM_008771488.1 | XP_008769710.1 | 4 | 348 | |||
| GenBank mRNA | BC079356.1 | - | 6 | 538 | ||
| BC091224.1 | - | 7 | 539 | 7128668 | ||

Faf2
Genomic Map