siRNA Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024145.2 | NP_001019316.1 | 17 | 2059 | ||
| GenBank mRNA | AB195278.1 | - | 17 | 2059 | ||
| AK076378.1 | - | 12 | 1545 | |||
| BC039947.1 | - | 17 | 1908 | |||
| BC046400.1 | - | 4 | 502 | |||
| BC145426.1 | - | 17 | 2011 | 9053984 | ||
| BC145427.1 | - | 17 | 2006 | |||
| BC145428.1 | - | 17 | 2011 | 9053986 | ||
| BC150771.1 | - | 17 | 2011 | 9087682 | ||
| DQ904008.1 | - | 17 | 2059 | |||

Pla2g4f
Genomic Map