siRNA Details
| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024234.1 | NP_001019405.1 | 1 | 654 | ||
| GenBank mRNA | BC094520.1 | - | 1 | 654 | 7316275 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wscd1| Gene Symbol: Wscd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024234.1 | NP_001019405.1 | 1 | 654 | ||
| GenBank mRNA | BC094520.1 | - | 1 | 654 | 7316275 | |