siRNA Details
| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024237.1 | NP_001019408.1 | 7 | 833 | ||
| GenBank mRNA | BC094515.1 | - | 7 | 833 | 7314191 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farsa| Gene Symbol: Farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024237.1 | NP_001019408.1 | 7 | 833 | ||
| GenBank mRNA | BC094515.1 | - | 7 | 833 | 7314191 | |