siRNA Details
| Gene Symbol: Eprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024238.1 | NP_001019409.1 | 6 | 490 | ||
| XM_006250425.2 | XP_006250487.1 | 5 | 590 | |||
| XM_006250426.2 | XP_006250488.1 | 5 | 590 | |||
| GenBank mRNA | AY383705.1 | - | 6 | 490 | ||

Eprs
Genomic Map