siRNA Details
| Gene Symbol: Eprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024238.1 | NP_001019409.1 | 27 | 3714 | ||
| XM_006250425.2 | XP_006250487.1 | 28 | 3967 | |||
| XM_006250426.2 | XP_006250488.1 | 27 | 3946 | |||
| XM_006250427.2 | XP_006250489.1 | 24 | 3347 | |||
| GenBank mRNA | AY383705.1 | - | 27 | 3714 | ||
| BC088324.1 | - | 7 | 766 | |||
| FQ220944.1 | - | 3 | 268 | |||

Eprs
Genomic Map