siRNA Details
| Gene Symbol: Ccdc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024245.1 | NP_001019416.1 | 3 | 884 | ||
| GenBank mRNA | BC093612.1 | - | 3 | 884 | 7307602 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc32| Gene Symbol: Ccdc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024245.1 | NP_001019416.1 | 3 | 884 | ||
| GenBank mRNA | BC093612.1 | - | 3 | 884 | 7307602 | |