siRNA Details
| Gene Symbol: Bop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024250.1 | NP_001019421.1 | 16 | 2192 | ||
| GenBank mRNA | BC092594.1 | - | 16 | 2192 | 7380064 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bop1| Gene Symbol: Bop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024250.1 | NP_001019421.1 | 16 | 2192 | ||
| GenBank mRNA | BC092594.1 | - | 16 | 2192 | 7380064 | |