siRNA Details
| Gene Symbol: Naa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024742.1 | NP_001019913.1 | 2 | 857 | ||
| XM_006250697.2 | XP_006250759.1 | 4 | 839 | |||
| GenBank mRNA | BC078912.1 | - | 2 | 830 | ||
| BC097350.1 | - | 2 | 857 | 7462336 | ||

Naa11
Genomic Map