siRNA Details
| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024744.1 | NP_001019915.1 | 6 | 622 | ||
| GenBank mRNA | BC097498.1 | - | 6 | 622 | 7462333 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc16| Gene Symbol: Cdc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024744.1 | NP_001019915.1 | 6 | 622 | ||
| GenBank mRNA | BC097498.1 | - | 6 | 622 | 7462333 | |