siRNA Details
| Gene Symbol: RGD1305713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024749.1 | NP_001019920.1 | 4 | 1562 | ||
| GenBank mRNA | BC094529.1 | - | 4 | 1562 | 7320095 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305713| Gene Symbol: RGD1305713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024749.1 | NP_001019920.1 | 4 | 1562 | ||
| GenBank mRNA | BC094529.1 | - | 4 | 1562 | 7320095 | |