siRNA Details
| Gene Symbol: Mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024865.1 | NP_001020036.1 | 4 | 1552 | ||
| GenBank mRNA | BC097472.1 | - | 4 | 1552 | 7457772 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl40| Gene Symbol: Mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024865.1 | NP_001020036.1 | 4 | 1552 | ||
| GenBank mRNA | BC097472.1 | - | 4 | 1552 | 7457772 | |