siRNA Details
| Gene Symbol: Vps37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024867.1 | NP_001020038.1 | 3 | 473 | ||
| XM_008771289.1 | XP_008769511.1 | 3 | 535 | |||
| GenBank mRNA | BC098069.1 | - | 3 | 473 | 7444656 | |
| FQ232149.1 | - | 3 | 527 | |||

Vps37a
Genomic Map