siRNA Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024869.1 | NP_001020040.1 | 3 | 652 | ||
| GenBank mRNA | BC097968.1 | - | 3 | 652 | 7441820 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wwtr1| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024869.1 | NP_001020040.1 | 3 | 652 | ||
| GenBank mRNA | BC097968.1 | - | 3 | 652 | 7441820 | |