siRNA Details
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024869.1 | NP_001020040.1 | 6, 7 | 1172 | ||
GenBank mRNA | BC097968.1 | - | 6, 7 | 1172 | 7441820 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024869.1 | NP_001020040.1 | 6, 7 | 1172 | ||
GenBank mRNA | BC097968.1 | - | 6, 7 | 1172 | 7441820 |