siRNA Details
| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024870.1 | NP_001020041.1 | 15, 16 | 1662 | ||
| GenBank mRNA | BC097352.1 | - | 15, 16 | 1662 | 7387542 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnnbl1| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024870.1 | NP_001020041.1 | 15, 16 | 1662 | ||
| GenBank mRNA | BC097352.1 | - | 15, 16 | 1662 | 7387542 | |