siRNA Details
| Gene Symbol: Dnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024879.1 | NP_001020050.1 | 15 | 1522 | ||
| XM_006245184.2 | XP_006245246.1 | 15 | 1520 | |||
| XM_006245185.2 | XP_006245247.1 | 15 | 1621 | |||
| XM_006245186.2 | XP_006245248.1 | 15 | 1573 | |||
| GenBank mRNA | BC081986.1 | - | 12 | 2001 | ||
| BC097388.1 | - | 15 | 1522 | 7453069 | ||
| FQ229070.1 | - | 12 | 1491 | |||
| FQ234491.1 | - | 15 | 1430 | |||

Dnpep
Genomic Map