siRNA Details
| Gene Symbol: Tns4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024881.1 | NP_001020052.1 | 13 | 2440 | ||
| GenBank mRNA | BC097378.1 | - | 13 | 2440 | 7445075 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tns4| Gene Symbol: Tns4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024881.1 | NP_001020052.1 | 13 | 2440 | ||
| GenBank mRNA | BC097378.1 | - | 13 | 2440 | 7445075 | |