siRNA Details
Gene Symbol: Tmem106a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024967.1 | NP_001020138.1 | 8 | 1285 | ||
XM_008767977.1 | XP_008766199.1 | 8 | 1443 | |||
XM_008767978.1 | XP_008766200.1 | 9 | 1186 | |||
XM_008767979.1 | XP_008766201.1 | 9 | 1174 | |||
GenBank mRNA | BC091170.1 | - | 8 | 1285 | 7131003 |
