siRNA Details
Gene Symbol: Sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024970.2 | NP_001020141.2 | 7 | 1275 | ||
GenBank mRNA | BC091401.1 | - | 4 | 1158 | 7324146 | |
CV077309.1 | - | 2 | 407 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024970.2 | NP_001020141.2 | 7 | 1275 | ||
GenBank mRNA | BC091401.1 | - | 4 | 1158 | 7324146 | |
CV077309.1 | - | 2 | 407 |