siRNA Details
| Gene Symbol: Arglu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024972.1 | NP_001020143.1 | 4 | 1372 | ||
| GenBank mRNA | BC091344.1 | - | 4 | 1372 | 7315932 | |
| BC127459.1 | - | 4 | 1332 | 7125115 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arglu1| Gene Symbol: Arglu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024972.1 | NP_001020143.1 | 4 | 1372 | ||
| GenBank mRNA | BC091344.1 | - | 4 | 1372 | 7315932 | |
| BC127459.1 | - | 4 | 1332 | 7125115 | ||