siRNA Details
| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 2 | 377 | ||
| GenBank mRNA | BC091268.1 | - | 2 | 377 | 7380414 | |
| FQ224897.1 | - | 2 | 412 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026120, BC092281, AI509964, BB656580, BC043335, and others.

RGD1311863
Genomic Map