siRNA Details
| Gene Symbol: RGD1311863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024974.1 | NP_001020145.1 | 7, 8 | 1125 | ||
| GenBank mRNA | BC091268.1 | - | 7, 8 | 1125 | 7380414 | |
| FQ223854.1 | - | 3, 4 | 395 | |||
| FQ224897.1 | - | 7, 8 | 1160 | |||

RGD1311863
Genomic Map