siRNA Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024982.1 | NP_001020153.1 | 11 | 761 | ||
| XM_006237022.2 | XP_006237084.1 | 11 | 1741 | |||
| XM_008763190.1 | XP_008761412.1 | 10 | 1738 | |||
| GenBank mRNA | BC091347.1 | - | 11 | 761 | 7316601 | |
| FQ213359.1 | - | 3 | 196 | |||

Cpne9
Genomic Map