siRNA Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 5 | 356 | ||
| XM_008768051.1 | XP_008766273.1 | 5 | 698 | |||
| GenBank mRNA | BC091314.1 | - | 5 | 412 | 7312354 | |
| BC092210.1 | - | 5 | 356 | 7311688 | ||
| EU327027.1 | - | 5 | 361 | |||
| EU327028.1 | - | 4 | 317 | |||
| EU327029.1 | - | 5 | 414 | |||
| EU327030.1 | - | 4 | 278 | |||
| EU327031.1 | - | 3 | 265 | |||
| FQ213000.1 | - | 5 | 344 | |||

Smarce1
Genomic Map