siRNA Details
| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 11 | 2021 | ||
| GenBank mRNA | BC091314.1 | - | 11 | 2071 | 7312354 | |
| BC092210.1 | - | 11 | 2021 | 7311688 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smarce1| Gene Symbol: Smarce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024993.1 | NP_001020164.1 | 11 | 2021 | ||
| GenBank mRNA | BC091314.1 | - | 11 | 2071 | 7312354 | |
| BC092210.1 | - | 11 | 2021 | 7311688 | ||